Product Details

SNP ID
rs148192841
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:236394487 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACAGATGGGGAGAGGCACTAAA[A/G]CTCCTGGTCACCAAGAGGGTATGTA
Phenotype
MIM: 606603
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
EDARADD PubMed Links
Additional Information
For this assay, SNP(s) [rs966365] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
EDARADD
Gene Name
EDAR associated death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080738.3 108 Intron NP_542776.1
NM_145861.2 108 Missense Mutation ACT,GCT T15A NP_665860.2

View Full Product Details