Product Details

SNP ID
rs148193368
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:11847284 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCCCCGCCCGAGGGCACCTCC[A/C]TCTCTCTGGGCTGGGCTGACTTCCC
Phenotype
MIM: 602726 MIM: 108780
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLCN6 PubMed Links

Gene Details

Gene
CLCN6
Gene Name
chloride voltage-gated channel 6
There are no transcripts associated with this gene.

Gene
NPPA
Gene Name
natriuretic peptide A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006172.3 378 Missense Mutation GAG,GAT E93D NP_006163.1
Gene
NPPA-AS1
Gene Name
NPPA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details