Product Details

SNP ID
rs148233924
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:201963632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAATAGGTAGCTTTGCAGTTTG[G/T]GGAGGGCTGTTTTCCATGATTGACT
Phenotype
MIM: 605057
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TIMM17A PubMed Links

Gene Details

Gene
TIMM17A
Gene Name
translocase of inner mitochondrial membrane 17 homolog A (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006335.2 243 Missense Mutation TGG,TGT W69C NP_006326.1

View Full Product Details