Product Details

SNP ID
rs148291612
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:153543851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGGAAGACACAGTACTCTTGGA[A/C]GTCCACCTCGTTGTCCCTGTTGCTG
Phenotype
MIM: 176992 MIM: 114210 MIM: 176991 MIM: 114110
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928034 PubMed Links

Gene Details

Gene
LOC101928034
Gene Name
uncharacterized LOC101928034
There are no transcripts associated with this gene.

Gene
S100A3
Gene Name
S100 calcium binding protein A3
There are no transcripts associated with this gene.

Gene
S100A4
Gene Name
S100 calcium binding protein A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002961.2 332 Missense Mutation GTC,TTC V72F NP_002952.1
NM_019554.2 332 Missense Mutation GTC,TTC V72F NP_062427.1
Gene
S100A5
Gene Name
S100 calcium binding protein A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002962.1 332 Intron NP_002953.2
XM_011509860.2 332 Intron XP_011508162.1
XM_017002029.1 332 Intron XP_016857518.1
XM_017002030.1 332 Intron XP_016857519.1
XM_017002031.1 332 Intron XP_016857520.1
XM_017002032.1 332 Intron XP_016857521.1
Gene
S100A6
Gene Name
S100 calcium binding protein A6
There are no transcripts associated with this gene.

View Full Product Details