Product Details

SNP ID
rs148351388
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:167054353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTGTCACTGGTCGAGGTGGTCC[A/G]GGGATTCACGCCCAGTGCTCCTCTG
Phenotype
MIM: 602171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPA33 PubMed Links

Gene Details

Gene
GPA33
Gene Name
glycoprotein A33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005814.2 1113 Missense Mutation NP_005805.1
XM_017000005.1 1113 Missense Mutation XP_016855494.1

View Full Product Details