Product Details

SNP ID
rs148375757
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:99712139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTAGCAGCTGTTCCCATACTCCA[A/G]TGAGTCCAGTTAAACATTTGCCTTC
Phenotype
MIM: 611578
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRRS1 PubMed Links

Gene Details

Gene
FRRS1
Gene Name
ferric chelate reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013660.2 1575 Silent Mutation CAC,CAT H482H NP_001013682.2
XM_005270861.3 1575 Silent Mutation CAC,CAT H482H XP_005270918.1
XM_011541451.2 1575 Silent Mutation CAC,CAT H482H XP_011539753.1
XM_011541452.1 1575 Silent Mutation CAC,CAT H482H XP_011539754.1
XM_017001271.1 1575 Silent Mutation CAC,CAT H482H XP_016856760.1

View Full Product Details