Product Details

SNP ID
rs148398789
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:228174812 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGAAGGTCACGGTGGAGCCGCA[C/T]CCGGAGCTGCGAGTGTGGGCGGTGT
Phenotype
MIM: 615316
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IBA57 PubMed Links

Gene Details

Gene
IBA57
Gene Name
IBA57 homolog, iron-sulfur cluster assembly
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010867.3 471 Silent Mutation CAC,CAT H154H NP_001010867.1
NM_001310327.1 471 UTR 5 NP_001297256.1
Gene
IBA57-AS1
Gene Name
IBA57 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details