Product Details

SNP ID
rs148410684
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:1071926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGTCTTGAGCGCGCACTGCACC[A/G]GCCAGAGTGCCACACAGAAGAGCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf159 PubMed Links

Gene Details

Gene
C1orf159
Gene Name
chromosome 1 open reading frame 159
There are no transcripts associated with this gene.

Gene
LOC100288175
Gene Name
uncharacterized LOC100288175
There are no transcripts associated with this gene.

Gene
LOC105378948
Gene Name
uncharacterized LOC105378948
There are no transcripts associated with this gene.

Gene
RNF223
Gene Name
ring finger protein 223
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205252.1 942 Missense Mutation CCG,CTG P214L NP_001192181.1
XM_017001286.1 942 Missense Mutation CCG,CTG P214L XP_016856775.1
XM_017001287.1 942 Intron XP_016856776.1

View Full Product Details