Product Details
- SNP ID
-
rs148415294
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:159535486 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGGTAAATGTCACAAAAGAAGTG[A/G]TCTACCACTGTGCCACAGAACGGCA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR10J5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4656837] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR10J5
- Gene Name
- olfactory receptor family 10 subfamily J member 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001004469.1 |
522 |
Silent Mutation |
GAC,GAT |
D174D |
NP_001004469.1 |
View Full Product Details