Product Details

SNP ID
rs148533540
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:158717696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAACCTGCAGAGAGTTGAGCACA[A/G]AGCCGGGGGGTCATGATCATTTGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6K3 PubMed Links

Gene Details

Gene
OR6K3
Gene Name
olfactory receptor family 6 subfamily K member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005327.2 420 Silent Mutation CTC,CTT L140L NP_001005327.2

View Full Product Details