Product Details

SNP ID
rs148544674
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:15573723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTAGCCGCCAGCAGGGCTGTGTT[A/C]CCTAAGAAAAAGTAAAACCTCACAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AGMAT PubMed Links

Gene Details

Gene
AGMAT
Gene Name
agmatinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024758.4 1130 Silent Mutation NP_079034.3
Gene
DNAJC16
Gene Name
DnaJ heat shock protein family (Hsp40) member C16
There are no transcripts associated with this gene.

View Full Product Details