Product Details

SNP ID
rs148583112
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:158547627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAAAAACCATCTTAATCATGGCA[A/G]TCATGAGTCCACAGAACCAGCATCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR6Y1 PubMed Links

Gene Details

Gene
OR6Y1
Gene Name
olfactory receptor family 6 subfamily Y member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005189.1 479 Missense Mutation ACT,ATT T160I NP_001005189.1

View Full Product Details