Product Details

SNP ID
rs148623746
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:45628296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCATCTGATGTTTCACTGCTACTG[C/G]TTTCGGTGTCTGAGTCCTCAAACTC
Phenotype
MIM: 603185
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC17 PubMed Links

Gene Details

Gene
CCDC17
Gene Name
coiled-coil domain containing 17
There are no transcripts associated with this gene.

Gene
GPBP1L1
Gene Name
GC-rich promoter binding protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021639.4 2648 Missense Mutation ACC,AGC T462S NP_067652.1
XM_017002019.1 2648 Missense Mutation ACC,AGC T409S XP_016857508.1
Gene
NASP
Gene Name
nuclear autoantigenic sperm protein
There are no transcripts associated with this gene.

View Full Product Details