Product Details
- SNP ID
-
rs148627056
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
5
- Location
-
Chr.1:155612511 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTCTTGTCTGCCCCTTGTCCTTG[C/G]GTGGGAGCCTGGGCCTGCGACCCGA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MSTO1
PubMed Links
Gene Details
- Gene
- MSTO1
- Gene Name
- misato 1, mitochondrial distribution and morphology regulator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256532.1 |
994 |
Missense Mutation |
CGT,GGT |
R303G |
NP_001243461.1 |
NM_001256533.1 |
994 |
Missense Mutation |
CGT,GGT |
R303G |
NP_001243462.1 |
NM_018116.3 |
994 |
Missense Mutation |
CGT,GGT |
R303G |
NP_060586.2 |
XM_006711421.1 |
994 |
Missense Mutation |
CGT,GGT |
R248G |
XP_006711484.1 |
XM_006711423.2 |
994 |
Missense Mutation |
CGT,GGT |
R125G |
XP_006711486.1 |
XM_011509695.1 |
994 |
Missense Mutation |
CGT,GGT |
R303G |
XP_011507997.1 |
XM_011509696.1 |
994 |
Missense Mutation |
CGT,GGT |
R303G |
XP_011507998.1 |
XM_011509697.1 |
994 |
Missense Mutation |
CGT,GGT |
R303G |
XP_011507999.1 |
XM_011509698.1 |
994 |
Missense Mutation |
CGT,GGT |
R303G |
XP_011508000.1 |
XM_011509699.1 |
994 |
Missense Mutation |
CGT,GGT |
R303G |
XP_011508001.1 |
XM_011509700.1 |
994 |
Missense Mutation |
CGT,GGT |
R303G |
XP_011508002.1 |
XM_011509701.1 |
994 |
Missense Mutation |
CGT,GGT |
R303G |
XP_011508003.1 |
XM_011509702.1 |
994 |
Intron |
|
|
XP_011508004.1 |
XM_011509703.1 |
994 |
Missense Mutation |
CGT,GGT |
R248G |
XP_011508005.1 |
XM_011509704.1 |
994 |
Missense Mutation |
CGT,GGT |
R248G |
XP_011508006.1 |
XM_011509705.1 |
994 |
Missense Mutation |
CGT,GGT |
R248G |
XP_011508007.1 |
XM_011509706.1 |
994 |
Missense Mutation |
CGT,GGT |
R126G |
XP_011508008.1 |
XM_011509707.1 |
994 |
Missense Mutation |
CGT,GGT |
R125G |
XP_011508009.1 |
XM_011509708.1 |
994 |
Missense Mutation |
CGT,GGT |
R125G |
XP_011508010.1 |
XM_011509709.2 |
994 |
Missense Mutation |
CGT,GGT |
R303G |
XP_011508011.1 |
XM_017001603.1 |
994 |
Missense Mutation |
CGT,GGT |
R126G |
XP_016857092.1 |
XM_017001604.1 |
994 |
Missense Mutation |
CGT,GGT |
R126G |
XP_016857093.1 |
XM_017001605.1 |
994 |
Missense Mutation |
CGT,GGT |
R126G |
XP_016857094.1 |
XM_017001606.1 |
994 |
Missense Mutation |
CGT,GGT |
R125G |
XP_016857095.1 |
XM_017001607.1 |
994 |
Missense Mutation |
CGT,GGT |
R125G |
XP_016857096.1 |
XM_017001608.1 |
994 |
Missense Mutation |
CGT,GGT |
R125G |
XP_016857097.1 |
XM_017001609.1 |
994 |
Missense Mutation |
CGT,GGT |
R125G |
XP_016857098.1 |
XM_017001610.1 |
994 |
Missense Mutation |
CGT,GGT |
R122G |
XP_016857099.1 |
XM_017001611.1 |
994 |
Missense Mutation |
CGT,GGT |
R122G |
XP_016857100.1 |
XM_017001612.1 |
994 |
Missense Mutation |
CGT,GGT |
R125G |
XP_016857101.1 |
XM_017001613.1 |
994 |
Missense Mutation |
CGT,GGT |
R122G |
XP_016857102.1 |
XM_017001614.1 |
994 |
Missense Mutation |
CGT,GGT |
R125G |
XP_016857103.1 |
XM_017001615.1 |
994 |
Missense Mutation |
CGT,GGT |
R122G |
XP_016857104.1 |
XM_017001616.1 |
994 |
Intron |
|
|
XP_016857105.1 |
View Full Product Details