Product Details

SNP ID
rs148654146
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:40532456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGAATGCTGAGTAAATTTCGAT[C/T]TGTAGTCATTTTGTAGAGTCTCTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF684 PubMed Links

Gene Details

Gene
ZNF684
Gene Name
zinc finger protein 684
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152373.3 870 Intron NP_689586.3
XM_011540671.2 870 Intron XP_011538973.1
XM_011540672.2 870 Intron XP_011538974.1
XM_011540673.2 870 Intron XP_011538975.1
XM_017000290.1 870 UTR 5 XP_016855779.1
XM_017000291.1 870 Intron XP_016855780.1
XM_017000292.1 870 Intron XP_016855781.1

View Full Product Details