Product Details

SNP ID
rs148695736
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:1212023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTGGGCTGGACAGTGATGGGCC[C/T]GGCCGGGGGGCCCTGGGTCTCCTGG
Phenotype
MIM: 614282 MIM: 603905 MIM: 600315
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDF4 PubMed Links

Gene Details

Gene
SDF4
Gene Name
stromal cell derived factor 4
There are no transcripts associated with this gene.

Gene
TNFRSF18
Gene Name
TNF receptor superfamily member 18
There are no transcripts associated with this gene.

Gene
TNFRSF4
Gene Name
TNF receptor superfamily member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003327.3 948 Missense Mutation AGG,GGG R185G NP_003318.1
XM_011542074.2 948 Missense Mutation AGG,GGG R189G XP_011540376.1
XM_011542075.2 948 Missense Mutation AGG,GGG R185G XP_011540377.1
XM_011542076.2 948 Missense Mutation AGG,GGG R189G XP_011540378.1
XM_011542077.2 948 Missense Mutation AGG,GGG R85G XP_011540379.1
XM_017002231.1 948 Missense Mutation AGG,GGG R185G XP_016857720.1
XM_017002232.1 948 Missense Mutation AGG,GGG R185G XP_016857721.1

View Full Product Details