Product Details

SNP ID
rs148711625
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:930204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACAGTCGCCCTGCCTGCCGCCC[A/G]GAACCTGAAGAAGGAGCGAACTCCC
Phenotype
MIM: 616765
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107985728 PubMed Links

Gene Details

Gene
LOC107985728
Gene Name
collagen alpha-1(III) chain-like
There are no transcripts associated with this gene.

Gene
LOC284600
Gene Name
uncharacterized LOC284600
There are no transcripts associated with this gene.

Gene
SAMD11
Gene Name
sterile alpha motif domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152486.2 202 Missense Mutation CAG,CGG Q41R NP_689699.2

View Full Product Details