Product Details

SNP ID
rs148734945
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:66814152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTATGCTGATTGGTTTGACTTG[A/G]ATCCAAGCAAAGTATCCATTATATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR78 PubMed Links

Gene Details

Gene
WDR78
Gene Name
WD repeat domain 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024763.4 2339 Missense Mutation TCC,TTC S842F NP_079039.4
NM_207014.2 2339 Intron NP_996897.2
XM_005271212.3 2339 Missense Mutation TCC,TTC S777F XP_005271269.1
XM_011542161.2 2339 Missense Mutation TCC,TTC S813F XP_011540463.1
XM_011542162.2 2339 Missense Mutation TCC,TTC S809F XP_011540464.1
XM_017002353.1 2339 Missense Mutation TCC,TTC S744F XP_016857842.1
XM_017002354.1 2339 Missense Mutation TCC,TTC S715F XP_016857843.1
XM_017002355.1 2339 Missense Mutation TCC,TTC S588F XP_016857844.1
XM_017002356.1 2339 Missense Mutation TCC,TTC S553F XP_016857845.1
XM_017002357.1 2339 Intron XP_016857846.1

View Full Product Details