Product Details

SNP ID
rs148842493
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:247037584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGAAGGGCACTGGAAGAAGTAAA[C/T]GACTTACCACATTCCTTACATCCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF670 PubMed Links

Gene Details

Gene
ZNF670
Gene Name
zinc finger protein 670
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204220.1 1252 Silent Mutation TCA,TCG S344S NP_001191149.1
NM_033213.4 1252 Silent Mutation TCA,TCG S345S NP_149990.1
Gene
ZNF670-ZNF695
Gene Name
ZNF670-ZNF695 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details