Product Details

SNP ID
rs148864594
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:151986210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTAAACATCATGGTTTCCATGGC[C/G]TGTTCCATTTGAGATGGCATTTTGG
Phenotype
MIM: 114085
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
S100A10 PubMed Links

Gene Details

Gene
S100A10
Gene Name
S100 calcium binding protein A10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002966.2 529 Silent Mutation CAC,CAG H7Q NP_002957.1

View Full Product Details