Product Details

SNP ID
rs148895370
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:155322113 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGCTCCTGCTCAGATCTTAGC[C/G]CCGATGAGTCCCCTGTCTCAGTCTA
Phenotype
MIM: 134629
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FDPS PubMed Links

Gene Details

Gene
FDPS
Gene Name
farnesyl diphosphate synthase
There are no transcripts associated with this gene.

Gene
LOC105371451
Gene Name
uncharacterized LOC105371451
There are no transcripts associated with this gene.

Gene
RUSC1
Gene Name
RUN and SH3 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105203.1 569 Missense Mutation CCC,GCC P114A NP_001098673.1
NM_001105204.1 569 Missense Mutation CCC,GCC P114A NP_001098674.1
NM_001105205.1 569 Intron NP_001098675.1
NM_001278227.1 569 Intron NP_001265156.1
NM_001278228.1 569 Intron NP_001265157.1
NM_001278229.1 569 Intron NP_001265158.1
NM_001278230.1 569 Intron NP_001265159.1
NM_014328.4 569 Intron NP_055143.2
XM_006711254.1 569 Missense Mutation CCC,GCC P114A XP_006711317.1
XM_006711256.1 569 Missense Mutation CCC,GCC P114A XP_006711319.1
XM_006711257.1 569 Intron XP_006711320.1
XM_017000891.1 569 Missense Mutation CCC,GCC P114A XP_016856380.1
XM_017000892.1 569 Intron XP_016856381.1
Gene
RUSC1-AS1
Gene Name
RUSC1 antisense RNA 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039517.1 569 Intron NP_001034606.1

View Full Product Details