Product Details

SNP ID
rs148954244
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:66820895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATGTGACCCAGATCTAATCAAT[A/C]ATCTGATTTCCCTGGCCACATTTCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WDR78 PubMed Links

Gene Details

Gene
WDR78
Gene Name
WD repeat domain 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024763.4 Intron NP_079039.4
NM_207014.2 Intron NP_996897.2
XM_005271212.3 Intron XP_005271269.1
XM_011542161.2 Intron XP_011540463.1
XM_011542162.2 Intron XP_011540464.1
XM_017002353.1 Intron XP_016857842.1
XM_017002354.1 Intron XP_016857843.1
XM_017002355.1 Intron XP_016857844.1
XM_017002356.1 Intron XP_016857845.1
XM_017002357.1 Intron XP_016857846.1

View Full Product Details