Product Details

SNP ID
rs148985659
Assay Type
Functionally Tested
NCBI dbSNP Submissions
4
Location
Chr.1:1426272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTCGGGGCCCCCGCTGCTGCCC[A/G]GCCGGCTGCTGGCCGGGCTCCTGCT
Phenotype
MIM: 611901
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ANKRD65 PubMed Links

Gene Details

Gene
ANKRD65
Gene Name
ankyrin repeat domain 65
There are no transcripts associated with this gene.

Gene
LOC102724312
Gene Name
uncharacterized LOC102724312
There are no transcripts associated with this gene.

Gene
LOC107985729
Gene Name
uncharacterized LOC107985729
There are no transcripts associated with this gene.

Gene
TMEM88B
Gene Name
transmembrane protein 88B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146685.1 145 Missense Mutation AGC,GGC S49G NP_001140157.1
Gene
VWA1
Gene Name
von Willebrand factor A domain containing 1
There are no transcripts associated with this gene.

View Full Product Details