Product Details

SNP ID
rs149075251
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:219173972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATAGCGCCTCTCTGATCTTCCTG[C/T]ATGGCTCAGGTGGATTTCAATTTTA
Phenotype
MIM: 616548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYPLAL1 PubMed Links

Gene Details

Gene
LYPLAL1
Gene Name
lysophospholipase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300769.1 83 Missense Mutation CAT,TAT H28Y NP_001287698.1
NM_001300770.1 83 Missense Mutation CAT,TAT H28Y NP_001287699.1
NM_001300771.1 83 Intron NP_001287700.1
NM_001300772.1 83 Intron NP_001287701.1
NM_138794.4 83 Missense Mutation CAT,TAT H28Y NP_620149.2
XM_006711162.3 83 Missense Mutation CAT,TAT H28Y XP_006711225.1
XM_011509182.2 83 Missense Mutation CAT,TAT H28Y XP_011507484.1
XM_011509183.2 83 UTR 5 XP_011507485.1
XM_017000271.1 83 Missense Mutation CAT,TAT H28Y XP_016855760.1
XM_017000272.1 83 UTR 5 XP_016855761.1
Gene
LYPLAL1-AS1
Gene Name
LYPLAL1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details