Product Details

SNP ID
rs149082475
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:11069593 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGAAGTCTGACTGGCTGTAGTC[A/G]TAAGGCGTAAACTCTTTTTCTGGTG
Phenotype
MIM: 605960 MIM: 182891
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOSC10 PubMed Links

Gene Details

Gene
EXOSC10
Gene Name
exosome component 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001998.2 2533 Silent Mutation TAA,TAG *818* NP_001001998.1
NM_002685.3 2533 Silent Mutation TAA,TAG *793* NP_002676.1
XM_005263475.2 2533 Nonsense Mutation TAA,TAG *823* XP_005263532.2
XM_017001494.1 2533 Silent Mutation TAA,TAG *407* XP_016856983.1
XM_017001495.1 2533 Silent Mutation TAA,TAG *402* XP_016856984.1
XM_017001496.1 2533 Silent Mutation TAA,TAG *391* XP_016856985.1
XM_017001497.1 2533 Silent Mutation TAA,TAG *386* XP_016856986.1
Gene
SRM
Gene Name
spermidine synthase
There are no transcripts associated with this gene.

View Full Product Details