Product Details

SNP ID
rs149177710
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:150509697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCCCCCTCCCCACCCCTATCCC[A/G]AAGCCTCCCCATGGATCACCCTGAC
Phenotype
MIM: 602201 MIM: 616092 MIM: 612805
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ECM1 PubMed Links

Gene Details

Gene
ECM1
Gene Name
extracellular matrix protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202858.1 357 Missense Mutation CAA,CGA Q53R NP_001189787.1
NM_004425.3 357 Missense Mutation CAA,CGA Q53R NP_004416.2
NM_022664.2 357 Missense Mutation CAA,CGA Q53R NP_073155.2
Gene
FALEC
Gene Name
focally amplified long non-coding RNA in epithelial cancer
There are no transcripts associated with this gene.

Gene
TARS2
Gene Name
threonyl-tRNA synthetase 2, mitochondrial (putative)
There are no transcripts associated with this gene.

View Full Product Details