Product Details
- SNP ID
-
rs149178480
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:15577826 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCCCCCATCAGAGGAACCAGCGA[C/T]TTCATCCAGCAGTCTTCAGCCAGGA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AGMAT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11544523] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGMAT
- Gene Name
- agmatinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024758.4 |
902 |
Silent Mutation |
AAA,AAG |
K253K |
NP_079034.3 |
- Gene
- DNAJC16
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C16
There are no transcripts associated with this gene.
View Full Product Details