Product Details

SNP ID
rs149202070
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:78628052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGATATGGTTGAAAGATGCAGCC[A/G]TCAGGGATGTACTATAACAATGGCT
Phenotype
MIM: 613975
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFI44L PubMed Links

Gene Details

Gene
IFI44L
Gene Name
interferon induced protein 44 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006820.3 175 Missense Mutation CAT,CGT H46R NP_006811.2
XM_005270391.3 175 Intron XP_005270448.1
XM_005270392.4 175 Intron XP_005270449.1
XM_005270393.4 175 Intron XP_005270450.1
XM_006710304.3 175 Intron XP_006710367.1
XM_011540539.2 175 Missense Mutation CAT,CGT H46R XP_011538841.1
XM_017000120.1 175 Intron XP_016855609.1

View Full Product Details