Product Details

SNP ID
rs149371901
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:154983491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAAGCCTGAAGTAGAAAGAGAC[A/G]GGATTTTGGTCCGGGGTGGAGAGCG
Phenotype
MIM: 116900 MIM: 610595 MIM: 600560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CKS1B PubMed Links

Gene Details

Gene
CKS1B
Gene Name
CDC28 protein kinase regulatory subunit 1B
There are no transcripts associated with this gene.

Gene
FLAD1
Gene Name
flavin adenine dinucleotide synthetase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184891.1 198 Intron NP_001171820.1
NM_001184892.1 198 Intron NP_001171821.1
NM_025207.4 198 UTR 5 NP_079483.3
NM_201398.2 198 Intron NP_958800.1
Gene
MIR4258
Gene Name
microRNA 4258
There are no transcripts associated with this gene.

Gene
SHC1
Gene Name
SHC adaptor protein 1
There are no transcripts associated with this gene.

View Full Product Details