Product Details

SNP ID
rs149402253
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:27394548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCAAGGGCATTGCCACACTGGC[C/T]GTGGTGCTTGGAGCCTTTGCCGCCT
Phenotype
MIM: 600241 MIM: 605875
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR3 PubMed Links

Gene Details

Gene
GPR3
Gene Name
G protein-coupled receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005281.3 849 Silent Mutation GCC,GCT A250A NP_005272.1
Gene
WASF2
Gene Name
WAS protein family member 2
There are no transcripts associated with this gene.

View Full Product Details