Product Details

SNP ID
rs149426833
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:234605685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGTTACAAACACCTGAACGTAA[A/G]GATTTAAAATAGTGTTCCTGTCAAT
Phenotype
MIM: 615332
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRF2BP2 PubMed Links

Gene Details

Gene
IRF2BP2
Gene Name
interferon regulatory factor 2 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077397.1 3247 UTR 3 NP_001070865.1
NM_182972.2 3247 UTR 3 NP_892017.2

View Full Product Details