Product Details

SNP ID
rs149449945
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:154959260 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGGCCAGGAAAGCCAGGGTCC[A/G]GACCAGGAAAGGGACTTGTGTTAGG
Phenotype
MIM: 606903 MIM: 600560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928120 PubMed Links

Gene Details

Gene
LOC101928120
Gene Name
uncharacterized LOC101928120
There are no transcripts associated with this gene.

Gene
PBXIP1
Gene Name
PBX homeobox interacting protein 1
There are no transcripts associated with this gene.

Gene
PYGO2
Gene Name
pygopus family PHD finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138300.3 946 Missense Mutation NP_612157.1
Gene
SHC1
Gene Name
SHC adaptor protein 1
There are no transcripts associated with this gene.

View Full Product Details