Product Details

SNP ID
rs149456951
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:27393994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGCGCTAGTGGTGGCCATCATC[A/G]TGGGCACTCCTGCCTTCCGTGCCCC
Phenotype
MIM: 600241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR3 PubMed Links

Gene Details

Gene
GPR3
Gene Name
G protein-coupled receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005281.3 295 Missense Mutation ATG,GTG M66V NP_005272.1

View Full Product Details