Product Details

SNP ID
rs149457096
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:78890870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGTTAGAAAGAAAATCTTTCCTT[A/G]GGTGCAGTGATATCACTCCTGAGAA
Phenotype
MIM: 616419
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRL4 PubMed Links

Gene Details

Gene
ADGRL4
Gene Name
adhesion G protein-coupled receptor L4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022159.3 2513 UTR 3 NP_071442.2

View Full Product Details