Product Details

SNP ID
rs149506075
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:44785159 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCAGCAGAGTGGTGAGAGGCAC[A/G]TACATGTCCGGGTCTCCCAGGGCTG
Phenotype
MIM: 607336 MIM: 600357
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BEST4 PubMed Links

Gene Details

Gene
BEST4
Gene Name
bestrophin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153274.2 910 Silent Mutation TAC,TAT Y287Y NP_695006.1
XM_017001020.1 910 Silent Mutation TAC,TAT Y287Y XP_016856509.1
XM_017001021.1 910 Silent Mutation TAC,TAT Y287Y XP_016856510.1
XM_017001022.1 910 Silent Mutation TAC,TAT Y287Y XP_016856511.1
XM_017001023.1 910 Silent Mutation TAC,TAT Y287Y XP_016856512.1
Gene
RPS8
Gene Name
ribosomal protein S8
There are no transcripts associated with this gene.

Gene
SNORD38A
Gene Name
small nucleolar RNA, C/D box 38A
There are no transcripts associated with this gene.

Gene
SNORD38B
Gene Name
small nucleolar RNA, C/D box 38B
There are no transcripts associated with this gene.

Gene
SNORD46
Gene Name
small nucleolar RNA, C/D box 46
There are no transcripts associated with this gene.

Gene
SNORD55
Gene Name
small nucleolar RNA, C/D box 55
There are no transcripts associated with this gene.

View Full Product Details