Product Details

SNP ID
rs149511442
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:110037959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACAAGTGGGCTGCAGAGCTCTC[A/G]GGTAACGCACAGACCTTTGTGTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STRIP1 PubMed Links

Gene Details

Gene
STRIP1
Gene Name
striatin interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270768.1 285 UTR 5 NP_001257697.1
NM_033088.3 285 Silent Mutation TCA,TCG S83S NP_149079.2
XM_006710995.2 285 Intron XP_006711058.1
XM_017002629.1 285 Silent Mutation TCA,TCG S83S XP_016858118.1
XM_017002630.1 285 Silent Mutation TCA,TCG S83S XP_016858119.1

View Full Product Details