Product Details

SNP ID
rs149659319
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:33013259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTGCGAACACGACATCGGGGGT[C/T]TGGGATGCATCGATGGCGGAGTGGA
Phenotype
MIM: 103020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AK2 PubMed Links

Gene Details

Gene
AK2
Gene Name
adenylate kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199199.1 630 Silent Mutation CAA,CAG Q206Q NP_001186128.1
NM_001319139.1 630 Silent Mutation CAA,CAG Q166Q NP_001306068.1
NM_001319140.1 630 Silent Mutation CAA,CAG Q166Q NP_001306069.1
NM_001319141.1 630 Silent Mutation CAA,CAG Q214Q NP_001306070.1
NM_001319142.1 630 Silent Mutation CAA,CAG Q172Q NP_001306071.1
NM_001319143.1 630 UTR 3 NP_001306072.1
NM_001625.3 630 Silent Mutation CAA,CAG Q214Q NP_001616.1
NM_013411.4 630 Silent Mutation CAA,CAG Q214Q NP_037543.1

View Full Product Details