Product Details

SNP ID
rs149664958
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:169854639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTGAAAGCAAAGCAGGAATAGG[C/T]TTCTGCTCCCCTGAGGTAACAGGTT
Phenotype
MIM: 608192
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf112 PubMed Links

Gene Details

Gene
C1orf112
Gene Name
chromosome 1 open reading frame 112
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320047.1 1971 Intron NP_001306976.1
NM_001320048.1 1971 Intron NP_001306977.1
NM_001320050.1 1971 Intron NP_001306979.1
NM_001320051.1 1971 Intron NP_001306980.1
NM_018186.3 1971 Intron NP_060656.2
XM_005245317.4 1971 Intron XP_005245374.1
XM_011509735.2 1971 Intron XP_011508037.1
XM_017001722.1 1971 Intron XP_016857211.1
XM_017001723.1 1971 Intron XP_016857212.1
XM_017001724.1 1971 Intron XP_016857213.1
XM_017001725.1 1971 Intron XP_016857214.1
Gene
SCYL3
Gene Name
SCY1 like pseudokinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020423.6 1971 Silent Mutation AAC,AAT N546N NP_065156.5
NM_181093.3 1971 Silent Mutation AAC,AAT N600N NP_851607.2
XM_006711465.1 1971 Silent Mutation XP_006711528.1
XM_011509801.1 1971 Silent Mutation XP_011508103.1
XM_011509802.1 1971 Silent Mutation XP_011508104.1
XM_011509803.1 1971 Silent Mutation XP_011508105.1
XM_017001862.1 1971 Silent Mutation XP_016857351.1
XM_017001863.1 1971 Silent Mutation XP_016857352.1
XM_017001864.1 1971 Silent Mutation XP_016857353.1
XM_017001865.1 1971 Silent Mutation XP_016857354.1

View Full Product Details