Product Details

SNP ID
rs149671274
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:247711974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCAAGGCATCTTTGATAGAGGTG[C/T]GGACGTGAAGGAAAACTGTGGACCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6F1 PubMed Links

Gene Details

Gene
OR6F1
Gene Name
olfactory receptor family 6 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005286.1 782 Missense Mutation CAC,CGC H261R NP_001005286.1

View Full Product Details