Product Details

SNP ID
rs149680474
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:116577369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGGGCTCCAGGCAAGTAGGGGA[A/G]TAGTTGAGGTGTGGCTCTTTGATCC
Phenotype
MIM: 153420 MIM: 603491
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD58 PubMed Links

Gene Details

Gene
CD58
Gene Name
CD58 molecule
There are no transcripts associated with this gene.

Gene
IGSF3
Gene Name
immunoglobulin superfamily member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007237.2 4218 Silent Mutation TAC,TAT Y1176Y NP_001007238.1
NM_001542.3 4218 Silent Mutation TAC,TAT Y1196Y NP_001533.2
XM_005270793.2 4218 Silent Mutation TAC,TAT Y1196Y XP_005270850.1
XM_005270794.4 4218 Silent Mutation TAC,TAT Y1181Y XP_005270851.1
XM_006710593.3 4218 Silent Mutation TAC,TAT Y1201Y XP_006710656.1
XM_011541315.1 4218 Silent Mutation TAC,TAT Y1196Y XP_011539617.1
XM_011541316.2 4218 Silent Mutation TAC,TAT Y639Y XP_011539618.1

View Full Product Details