Product Details

SNP ID
rs149700508
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:25301001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGTGTTCGCAGCCTATTTTGGGC[C/T]GTCTGTGGCCTGGTGCCTGCCAAAG
Phenotype
MIM: 111680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RHD PubMed Links

Gene Details

Gene
RHD
Gene Name
Rh blood group D antigen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127691.2 600 Missense Mutation CCG,CTG P181L NP_001121163.1
NM_001282867.1 600 Missense Mutation CCG,CTG P15L NP_001269796.1
NM_001282868.1 600 Missense Mutation CCG,CTG P181L NP_001269797.1
NM_001282869.1 600 Missense Mutation CCG,CTG P181L NP_001269798.1
NM_001282870.1 600 Missense Mutation CCG,CTG P181L NP_001269799.1
NM_001282871.1 600 Missense Mutation CCG,CTG P181L NP_001269800.1
NM_001282872.1 600 Missense Mutation CCG,CTG P181L NP_001269801.1
NM_016124.4 600 Missense Mutation CCG,CTG P181L NP_057208.2
XM_017002015.1 600 Intron XP_016857504.1
Gene
RSRP1
Gene Name
arginine and serine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321772.1 600 Intron NP_001308701.1
NM_020317.4 600 Intron NP_064713.3
XM_011541797.1 600 Intron XP_011540099.1

View Full Product Details