Product Details

SNP ID
rs149711133
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:156154662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGCTTCAGCTGCTGCTGCCGAC[A/G]ACGACCGCGGGGGGAGGCGGGCAGG
Phenotype
MIM: 607292
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEMA4A PubMed Links

Gene Details

Gene
SEMA4A
Gene Name
semaphorin 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193300.1 382 Silent Mutation ACA,ACG T28T NP_001180229.1
NM_001193301.1 382 Silent Mutation ACA,ACG T28T NP_001180230.1
NM_001193302.1 382 Intron NP_001180231.1
NM_022367.3 382 Silent Mutation ACA,ACG T28T NP_071762.2
XM_011509871.2 382 Intron XP_011508173.1
XM_011509872.2 382 Silent Mutation ACA,ACG T28T XP_011508174.1
XM_011509873.2 382 Silent Mutation ACA,ACG T28T XP_011508175.1
XM_011509874.2 382 Intron XP_011508176.1
XM_011509875.2 382 Intron XP_011508177.1
XM_011509876.2 382 Intron XP_011508178.1
XM_011509878.2 382 Intron XP_011508180.1
XM_011509879.2 382 Intron XP_011508181.1
XM_017002056.1 382 Silent Mutation ACA,ACG T28T XP_016857545.1
XM_017002057.1 382 UTR 5 XP_016857546.1

View Full Product Details