Product Details

SNP ID
rs149772863
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:11857663 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCAGGACTTCCTCTTAATGCC[A/G]CCTCAGCACTGTCAGGGAAAGAGAG
Phenotype
MIM: 108780 MIM: 600295
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPPA PubMed Links

Gene Details

Gene
NPPA
Gene Name
natriuretic peptide A
There are no transcripts associated with this gene.

Gene
NPPB
Gene Name
natriuretic peptide B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002521.2 499 Missense Mutation CGG,TGG R133W NP_002512.1

View Full Product Details