Product Details

SNP ID
rs149812347
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:26863695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCATGGACATCAGCAAGAAGGA[C/G]ATGCCGCCCACCAACCCCATCCGCC
Phenotype
MIM: 601290
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SFN PubMed Links

Gene Details

Gene
SFN
Gene Name
stratifin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006142.3 554 Missense Mutation GAC,GAG D161E NP_006133.1
Gene
ZDHHC18
Gene Name
zinc finger DHHC-type containing 18
There are no transcripts associated with this gene.

View Full Product Details