Product Details

SNP ID
rs149831242
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:8324578 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGGACCTGCACCCCCAGAGGTC[C/T]TTCCGGGAGCTGCTTTTCAACGGCT
Phenotype
MIM: 605763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC45A1 PubMed Links

Gene Details

Gene
SLC45A1
Gene Name
solute carrier family 45 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080397.2 375 Silent Mutation TCC,TCT S117S NP_001073866.2
XM_011541530.1 375 Silent Mutation TCC,TCT S117S XP_011539832.1
XM_011541531.1 375 Silent Mutation TCC,TCT S117S XP_011539833.1

View Full Product Details