Product Details

SNP ID
rs149849664
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:1437129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGTTCCCCTTCGGCCAGCACAG[C/T]TCGGGTGAGGCTGCCCAGGATGCGG
Phenotype
MIM: 611901
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102724312 PubMed Links

Gene Details

Gene
LOC102724312
Gene Name
uncharacterized LOC102724312
There are no transcripts associated with this gene.

Gene
LOC107985729
Gene Name
uncharacterized LOC107985729
There are no transcripts associated with this gene.

Gene
TMEM88B
Gene Name
transmembrane protein 88B
There are no transcripts associated with this gene.

Gene
VWA1
Gene Name
von Willebrand factor A domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022834.4 502 Silent Mutation AGC,AGT S92S NP_073745.2
NM_199121.2 502 Intron NP_954572.2

View Full Product Details