Product Details

SNP ID
rs149856753
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:27100394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGCAGCGCTGTATCCTCTGGGA[A/G]CTGGGGCTGTCACTGGGCGCGGGGG
Phenotype
MIM: 107310
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC9A1 PubMed Links

Gene Details

Gene
SLC9A1
Gene Name
solute carrier family 9 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003047.4 2235 Silent Mutation AGC,AGT S787S NP_003038.2
XM_011542021.2 2235 Silent Mutation AGC,AGT S677S XP_011540323.1

View Full Product Details