Product Details

SNP ID
rs149954582
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:29148647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGCTGGATTCAGACTTGGCATG[G/T]TCCCGCTCCTTTGACACGGAGCGGG
Phenotype
MIM: 601940
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SRSF4 PubMed Links

Gene Details

Gene
SRSF4
Gene Name
serine and arginine rich splicing factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005626.4 1331 Silent Mutation NP_005617.2
XM_011541951.2 1331 Silent Mutation XP_011540253.1

View Full Product Details