Product Details

SNP ID
rs149959295
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:197039385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCTGCCATTCATTTCTATGTT[A/C]TTAAGGGTTCTTGATAAGACAGAGT
Phenotype
MIM: 134580
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
F13B PubMed Links

Gene Details

Gene
F13B
Gene Name
coagulation factor XIII B chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001994.2 2983 Missense Mutation AGA,ATA R660I NP_001985.2
XM_011509283.2 2983 UTR 3 XP_011507585.1
XM_011509284.2 2983 UTR 3 XP_011507586.1
XM_011509286.2 2983 UTR 3 XP_011507588.1

View Full Product Details