Product Details

SNP ID
rs149966116
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:46610084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTTGTAGCAGGTGTTGACGTGC[A/G]CCTCTGCCCCCATGCTGAGGATGCT
Phenotype
MIM: 606724
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MKNK1 PubMed Links

Gene Details

Gene
MKNK1
Gene Name
MAP kinase interacting serine/threonine kinase 1
There are no transcripts associated with this gene.

Gene
MOB3C
Gene Name
MOB kinase activator 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145279.4 3701 Missense Mutation GCG,GTG A232V NP_660322.2
NM_201403.2 3701 Missense Mutation GCG,GTG A180V NP_958805.1
XM_017000407.1 3701 Missense Mutation GCG,GTG A203V XP_016855896.1

View Full Product Details